Assay Details
Gene Symbol: GR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115323.4 | NP_191026.1 | 9 - 10 | 1764 | 76 | |
GenBank mRNA | BT020536.1 | - | 9 - 10 | 1670 | 76 | |
BT030335.1 | - | 9 - 10 | 1653 | 76 | ||
D14049.1 | - | 9 - 10 | 1671 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
