Assay Details
| Gene Symbol: CDT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115328.4 | NP_191031.1 | 2 - 3 | 343 | 107 | |
| GenBank mRNA | AJ421409.1 | - | 2 - 3 | 270 | 107 | |
| BT029185.1 | - | 2 - 3 | 270 | 107 | ||
More Information
Set Membership: |
|

CDT1B
Genomic Map