Assay Details
Gene Symbol: CDT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115328.4 | NP_191031.1 | 4 - 5 | 1199 | 158 | |
GenBank mRNA | BT029185.1 | - | 4 - 5 | 1126 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
