Assay Details
Gene Symbol: PIP2;5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115339.3 | NP_191042.1 | 3 - 4 | 1103 | 113 | |
GenBank mRNA | AF370351.1 | - | 3 - 4 | 840 | 113 | |
AY062981.1 | - | 3 - 4 | 745 | 113 | ||
AY084843.1 | - | 3 - 4 | 842 | 113 | ||
BT000670.1 | - | 3 - 4 | 761 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
