Assay Details
| Gene Symbol: MRH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339702.1 | NP_001326426.1 | 19 - 20 | 3015 | 82 | |
| NM_001339703.1 | NP_001326427.1 | 18 - 19 | 3121 | 82 | ||
| NM_115344.4 | NP_191047.3 | 18 - 19 | 3036 | 82 | ||
| GenBank mRNA | AB290928.1 | - | 19 - 20 | 2904 | 82 | |
| EU257694.1 | - | 19 - 20 | 2889 | 82 | ||
More Information
Set Membership: |
|

MRH2
Genomic Map