Assay Details
| Gene Symbol: PDIL1-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115353.5 | NP_191056.2 | 11 - 12 | 1627 | 63 | |
| GenBank mRNA | AJ271376.1 | - | 11 - 12 | 1651 | 63 | |
| AY093115.1 | - | 11 - 12 | 1593 | 63 | ||
| BT000172.1 | - | 11 - 12 | 1575 | 63 | ||
More Information
Set Membership: |
|

PDIL1-3
Genomic Map