Assay Details
| Gene Symbol: GSTL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115362.2 | NP_191064.1 | 3 - 4 | 349 | 92 | |
| GenBank mRNA | BT030006.1 | - | 3 - 4 | 308 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSTL2| Gene Symbol: GSTL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115362.2 | NP_191064.1 | 3 - 4 | 349 | 92 | |
| GenBank mRNA | BT030006.1 | - | 3 - 4 | 308 | 92 | |
Set Membership: |
|