Assay Details
Gene Symbol: OBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115395.6 | NP_191097.3 | 1 - 2 | 190 | 120 | |
NM_202713.3 | NP_974442.1 | 1 - 2 | 190 | 120 | ||
GenBank mRNA | AB493653.1 | - | 1 - 2 | 55 | 120 | |
AK221402.1 | - | 1 - 2 | 188 | 120 | ||
BT033028.1 | - | 1 - 2 | 55 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
