Assay Details
Gene Symbol: UBC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203167.1 | NP_001190096.1 | 1 - 2 | 152 | 80 | |
NM_115396.4 | NP_567020.1 | 1 - 2 | 490 | 80 | ||
GenBank mRNA | AK317143.1 | - | 1 - 2 | 64 | 80 | |
AY045883.1 | - | 1 - 2 | 148 | 80 | ||
AY086490.1 | - | 1 - 2 | 64 | 80 | ||
AY117262.1 | - | 1 - 2 | 39 | 80 | ||
DQ027028.1 | - | 1 - 2 | 39 | 80 | ||
X72625.1 | - | 134 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
