Assay Details
| Gene Symbol: UBC14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203167.1 | NP_001190096.1 | 2 - 3 | 265 | 74 | |
| NM_115396.4 | NP_567020.1 | 2 - 3 | 603 | 74 | ||
| GenBank mRNA | AK317143.1 | - | 2 - 3 | 177 | 74 | |
| AY045883.1 | - | 2 - 3 | 261 | 74 | ||
| AY086490.1 | - | 2 - 3 | 177 | 74 | ||
| AY117262.1 | - | 2 - 3 | 152 | 74 | ||
| DQ027028.1 | - | 2 - 3 | 152 | 74 | ||
| X72625.1 | - | 247 | 74 | |||
More Information
Set Membership: |
|

UBC14
Genomic Map