Assay Details
Gene Symbol: UBC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203167.1 | NP_001190096.1 | 3 - 4 | 351 | 118 | |
NM_115396.4 | NP_567020.1 | 3 - 4 | 689 | 118 | ||
GenBank mRNA | AK317143.1 | - | 3 - 4 | 263 | 118 | |
AY045883.1 | - | 3 - 4 | 347 | 118 | ||
AY086490.1 | - | 3 - 4 | 263 | 118 | ||
AY117262.1 | - | 3 - 4 | 238 | 118 | ||
DQ027028.1 | - | 3 - 4 | 238 | 118 | ||
X72625.1 | - | 333 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
