Assay Details
Gene Symbol: AT3G55390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115397.3 | NP_191099.1 | 1 - 2 | 588 | 61 | |
GenBank mRNA | AY088394.1 | - | 1 - 2 | 380 | 61 | |
BT024913.1 | - | 1 - 2 | 215 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
