Assay Details
Gene Symbol: AT3G55390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115397.3 | NP_191099.1 | 2 - 3 | 730 | 65 | |
GenBank mRNA | AY088394.1 | - | 2 - 3 | 522 | 65 | |
BT024913.1 | - | 2 - 3 | 357 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
