Assay Details
| Gene Symbol: TPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115402.4 | NP_191104.1 | 2 - 3 | 508 | 101 | |
| GenBank mRNA | AF375426.1 | - | 2 - 3 | 211 | 101 | |
| AY074822.1 | - | 2 - 3 | 117 | 101 | ||
More Information
Set Membership: |
|

TPI
Genomic Map