Assay Details
| Gene Symbol: TPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115402.4 | NP_191104.1 | 5 - 6 | 845 | 64 | |
| GenBank mRNA | AF375426.1 | - | 5 - 6 | 548 | 64 | |
| AY074822.1 | - | 5 - 6 | 454 | 64 | ||
More Information
Set Membership: |
|

TPI
Genomic Map