Assay Details
| Gene Symbol: TPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115402.4 | NP_191104.1 | 6 - 7 | 938 | 73 | |
| GenBank mRNA | AF375426.1 | - | 6 - 7 | 641 | 73 | |
| AY074822.1 | - | 6 - 7 | 547 | 73 | ||
More Information
Set Membership: |
|

TPI
Genomic Map