Assay Details
Gene Symbol: ROPGEF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339743.1 | NP_001325478.1 | 3 - 4 | 745 | 85 | |
NM_115424.3 | NP_191125.1 | 3 - 4 | 786 | 85 | ||
GenBank mRNA | BT020279.1 | - | 3 - 4 | 669 | 85 | |
BT021117.1 | - | 3 - 4 | 592 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
