Assay Details
Gene Symbol: LAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339751.1 | NP_001326017.1 | 3 - 4 | 426 | 115 | |
NM_115443.4 | NP_567027.2 | 3 - 4 | 432 | 115 | ||
NM_202716.3 | NP_974445.2 | 4 - 5 | 645 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
