Assay Details
| Gene Symbol: LAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339748.1 | NP_001319763.1 | 4 - 5 | 1131 | 79 | |
| NM_001339749.1 | NP_001326016.1 | 3 - 4 | 1320 | 79 | ||
| NM_001339750.1 | NP_001326018.1 | 3 - 4 | 1320 | 79 | ||
| NM_001339751.1 | NP_001326017.1 | 5 - 6 | 609 | 79 | ||
| NM_115443.4 | NP_567027.2 | 5 - 6 | 615 | 79 | ||
| NM_202716.3 | NP_974445.2 | 6 - 7 | 828 | 79 | ||
More Information
Set Membership: |
|

LAF3
Genomic Map