Assay Details
Gene Symbol: LAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339748.1 | NP_001319763.1 | 5 - 6 | 1256 | 71 | |
NM_001339749.1 | NP_001326016.1 | 4 - 5 | 1445 | 71 | ||
NM_001339750.1 | NP_001326018.1 | 4 - 5 | 1445 | 71 | ||
NM_001339751.1 | NP_001326017.1 | 6 - 7 | 734 | 71 | ||
NM_115443.4 | NP_567027.2 | 6 - 7 | 740 | 71 | ||
NM_202716.3 | NP_974445.2 | 7 - 8 | 953 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
