Assay Details
Gene Symbol: FER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115467.5 | NP_191168.1 | 1 - 2 | 355 | 105 | |
GenBank mRNA | AJ312190.1 | - | 1 - 2 | 347 | 105 | |
AK226273.1 | - | 1 - 2 | 353 | 105 | ||
AY072221.1 | - | 1 - 2 | 352 | 105 | ||
AY122951.1 | - | 1 - 2 | 304 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
