Assay Details
Gene Symbol: FER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115467.5 | NP_191168.1 | 2 - 3 | 444 | 61 | |
GenBank mRNA | AJ312190.1 | - | 2 - 3 | 436 | 61 | |
AK226273.1 | - | 2 - 3 | 442 | 61 | ||
AY072221.1 | - | 2 - 3 | 441 | 61 | ||
AY122951.1 | - | 2 - 3 | 393 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
