Assay Details
| Gene Symbol: AT3G56200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115478.7 | NP_191179.1 | 3 - 4 | 1048 | 80 | |
| GenBank mRNA | AY094388.1 | - | 3 - 4 | 762 | 80 | |
More Information
Set Membership: |
|

AT3G56200
Genomic Map