Assay Details
| Gene Symbol: ARR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339837.1 | NP_001325622.1 | 5 - 6 | 758 | 146 | |
| NM_115563.3 | NP_191263.1 | 4 - 5 | 1002 | 146 | ||
| GenBank mRNA | AB010918.1 | - | 4 - 5 | 561 | 146 | |
| AY090234.1 | - | 4 - 5 | 804 | 146 | ||
| AY133571.1 | - | 4 - 5 | 357 | 146 | ||
More Information
Set Membership: |
|

ARR9
Genomic Map