Assay Details
| Gene Symbol: ADPG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115611.2 | NP_191310.1 | 5 - 6 | 884 | 114 | |
| GenBank mRNA | AK117942.1 | - | 5 - 6 | 884 | 114 | |
| BT005376.1 | - | 5 - 6 | 785 | 114 | ||
More Information
Set Membership: |
|

ADPG1
Genomic Map