Assay Details
| Gene Symbol: SCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115649.3 | NP_191346.1 | 1 - 2 | 353 | 61 | |
| GenBank mRNA | AY042838.1 | - | 1 - 2 | 133 | 61 | |
| BT003377.1 | - | 1 - 2 | 65 | 61 | ||
| U44976.1 | - | 1 - 2 | 112 | 61 | ||
More Information
Set Membership: |
|

SCE1
Genomic Map