Assay Details
| Gene Symbol: SCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115649.3 | NP_191346.1 | 3 - 4 | 596 | 75 | |
| GenBank mRNA | AY042838.1 | - | 3 - 4 | 376 | 75 | |
| BT003377.1 | - | 3 - 4 | 308 | 75 | ||
| U44976.1 | - | 3 - 4 | 355 | 75 | ||
More Information
Set Membership: |
|

SCE1
Genomic Map