Assay Details
Gene Symbol: AGAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036778.2 | NP_001031855.1 | 2 - 3 | 164 | 87 | |
NM_120921.5 | NP_568193.1 | 1 - 2 | 277 | 87 | ||
GenBank mRNA | AK317201.1 | - | 2 - 3 | 165 | 87 | |
AY085529.1 | - | 1 - 2 | 193 | 87 | ||
AY090238.1 | - | 1 - 2 | 144 | 87 | ||
BT000619.1 | - | 1 - 2 | 133 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
