Assay Details
Gene Symbol: AGAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036778.2 | NP_001031855.1 | 8 - 9 | 618 | 77 | |
NM_120921.5 | NP_568193.1 | 7 - 8 | 731 | 77 | ||
GenBank mRNA | AK317201.1 | - | 8 - 9 | 619 | 77 | |
AY085529.1 | - | 7 - 8 | 647 | 77 | ||
AY090238.1 | - | 7 - 8 | 598 | 77 | ||
BT000619.1 | - | 7 - 8 | 587 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
