Assay Details
Gene Symbol: AGAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036778.2 | NP_001031855.1 | 10 - 11 | 766 | 83 | |
NM_120921.5 | NP_568193.1 | 9 - 10 | 879 | 83 | ||
GenBank mRNA | AK317201.1 | - | 10 - 11 | 767 | 83 | |
AY085529.1 | - | 9 - 10 | 795 | 83 | ||
AY090238.1 | - | 9 - 10 | 746 | 83 | ||
BT000619.1 | - | 9 - 10 | 735 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
