Assay Details
Gene Symbol: AGAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342998.1 | NP_001332077.1 | 10 - 11 | 880 | 67 | |
NM_120922.5 | NP_196455.1 | 10 - 11 | 937 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
