Assay Details
| Gene Symbol: CI51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120938.4 | NP_196470.1 | 2 - 3 | 1774 | 97 | |
| GenBank mRNA | AY092971.1 | - | 2 - 3 | 1590 | 97 | |
More Information
Set Membership: |
|

CI51
Genomic Map