Assay Details
| Gene Symbol: AT5G08540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120940.3 | NP_568200.1 | 2 - 3 | 639 | 92 | |
| GenBank mRNA | AY035084.1 | - | 2 - 3 | 639 | 92 | |
| AY051035.1 | - | 2 - 3 | 548 | 92 | ||
More Information
Set Membership: |
|

AT5G08540
Genomic Map