Assay Details
Gene Symbol: SNRK2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343022.1 | NP_001331206.1 | 6 - 7 | 1243 | 90 | |
NM_120946.5 | NP_196476.1 | 7 - 8 | 1011 | 90 | ||
GenBank mRNA | BT024886.1 | - | 7 - 8 | 642 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
