Assay Details
| Gene Symbol: EMB2107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125730.1 | NP_001119202.1 | 10 - 11 | 1273 | 75 | |
| NM_121027.4 | NP_196552.1 | 10 - 11 | 1694 | 75 | ||
| NM_203029.2 | NP_974758.1 | 10 - 11 | 1240 | 75 | ||
| GenBank mRNA | AX765868.1 | - | 9 - 10 | 1108 | 75 | |
| AY034908.1 | - | 10 - 11 | 1241 | 75 | ||
| AY113906.1 | - | 9 - 10 | 1108 | 75 | ||
| AY230832.1 | - | 9 - 10 | 1114 | 75 | ||
More Information
Set Membership: |
|

EMB2107
Genomic Map