Assay Details
| Gene Symbol: EMB2107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125730.1 | NP_001119202.1 | 6 - 7 | 823 | 86 | |
| NM_121027.4 | NP_196552.1 | 6 - 7 | 1248 | 86 | ||
| NM_203029.2 | NP_974758.1 | 6 - 7 | 794 | 86 | ||
| GenBank mRNA | AX765868.1 | - | 5 - 6 | 662 | 86 | |
| AY034908.1 | - | 6 - 7 | 795 | 86 | ||
| AY113906.1 | - | 5 - 6 | 662 | 86 | ||
| AY230832.1 | - | 5 - 6 | 668 | 86 | ||
More Information
Set Membership: |
|

EMB2107
Genomic Map