Assay Details
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036806.2 | NP_001031883.1 | 4 - 5 | 825 | 88 | |
NM_001036807.1 | NP_001031884.1 | 4 - 5 | 709 | 88 | ||
NM_001085114.1 | NP_001078583.1 | 4 - 5 | 709 | 88 | ||
NM_001343359.1 | NP_001330235.1 | 4 - 5 | 1982 | 88 | ||
NM_001343360.1 | NP_001330236.1 | 4 - 5 | 1587 | 88 | ||
NM_001343361.1 | NP_001330237.1 | 4 - 5 | 1055 | 88 | ||
NM_001343362.1 | NP_001330238.1 | 3 - 4 | 512 | 88 | ||
NM_121478.4 | NP_568303.2 | 4 - 5 | 747 | 88 | ||
NM_203053.2 | NP_974782.1 | 4 - 5 | 714 | 88 | ||
GenBank mRNA | AF324712.2 | - | 3 - 4 | 304 | 88 | |
AF326863.1 | - | 3 - 4 | 304 | 88 | ||
AF339686.1 | - | 2 - 3 | 208 | 88 | ||
AF428428.1 | - | 3 - 4 | 345 | 88 | ||
BT000652.1 | - | 3 - 4 | 302 | 88 | ||
BT000663.1 | - | 3 - 4 | 306 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
