Assay Details
| Gene Symbol: ABIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343870.1 | NP_001318635.1 | 5 - 6 | 746 | 79 | |
| NM_001343871.1 | NP_001330711.1 | 5 - 6 | 767 | 79 | ||
| NM_001343872.1 | NP_001330710.1 | 5 - 6 | 706 | 79 | ||
| NM_122337.5 | NP_197819.2 | 5 - 6 | 782 | 79 | ||
| GenBank mRNA | AK222217.1 | - | 5 - 6 | 704 | 79 | |
| AK228814.1 | - | 5 - 6 | 584 | 79 | ||
| AY817014.1 | - | 4 - 5 | 432 | 79 | ||
| BT011734.1 | - | 4 - 5 | 432 | 79 | ||
More Information
Set Membership: |
|

ABIL3
Genomic Map