Assay Details
Gene Symbol: DES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344053.1 | NP_001330586.1 | 5 - 6 | 889 | 81 | |
NM_001344054.1 | NP_001330588.1 | 5 - 6 | 908 | 81 | ||
NM_001344055.1 | NP_001330587.1 | 5 - 6 | 810 | 81 | ||
NM_122686.2 | NP_198155.1 | 4 - 5 | 770 | 81 | ||
NM_203114.2 | NP_974843.1 | 5 - 6 | 958 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
