Assay Details
Gene Symbol: BON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125583.4 | NP_568944.1 | 9 - 10 | 924 | 85 | |
NM_203248.2 | NP_974977.1 | 8 - 9 | 959 | 85 | ||
GenBank mRNA | AX380787.1 | - | 8 - 9 | 881 | 85 | |
AY062824.1 | - | 9 - 10 | 915 | 85 | ||
AY085339.1 | - | 9 - 10 | 916 | 85 | ||
BT010555.1 | - | 8 - 9 | 837 | 85 |
Gene Symbol: AT5G61910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203661.1 | NP_001190590.1 | 15 - 16 | 3360 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
