Assay Details
| Gene Symbol: FAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125859.4 | NP_568990.1 | 7 - 8 | 878 | 70 | |
| NM_203262.3 | NP_974991.1 | 7 - 8 | 878 | 70 | ||
| NM_203263.2 | NP_974992.1 | 7 - 8 | 793 | 70 | ||
| GenBank mRNA | AF375435.1 | - | 7 - 8 | 814 | 70 | |
| AY074827.1 | - | 7 - 8 | 745 | 70 | ||
More Information
Set Membership: |
|

FAS2
Genomic Map