Assay Details
| Gene Symbol: FAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125859.4 | NP_568990.1 | 8 - 9 | 951 | 76 | |
| NM_203262.3 | NP_974991.1 | 8 - 9 | 951 | 76 | ||
| NM_203263.2 | NP_974992.1 | 8 - 9 | 866 | 76 | ||
| GenBank mRNA | AF375435.1 | - | 8 - 9 | 887 | 76 | |
| AY074827.1 | - | 8 - 9 | 818 | 76 | ||
More Information
Set Membership: |
|

FAS2
Genomic Map