Assay Details
| Gene Symbol: HAF01 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103009.4 | NP_174552.1 | 3 - 4 | 447 | 69 | |
| GenBank mRNA | AF510669.1 | - | 2 - 3 | 155 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HAF01| Gene Symbol: HAF01 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103009.4 | NP_174552.1 | 3 - 4 | 447 | 69 | |
| GenBank mRNA | AF510669.1 | - | 2 - 3 | 155 | 69 | |
Set Membership: |
|