Assay Details
| Gene Symbol: CID11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036054.2 | NP_001031131.1 | 6 - 7 | 1041 | 96 | |
| NM_103013.3 | NP_174556.1 | 6 - 7 | 1203 | 96 | ||
More Information
Set Membership: |
|

CID11
Genomic Map