Assay Details
| Gene Symbol: CID11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103013.3 | NP_174556.1 | 9 - 10 | 1382 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CID11| Gene Symbol: CID11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103013.3 | NP_174556.1 | 9 - 10 | 1382 | 74 | |
Set Membership: |
|