Assay Details
| Gene Symbol: AT1G33360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103063.4 | NP_564423.3 | 5 - 6 | 1420 | 74 | |
| GenBank mRNA | BT015363.1 | - | 5 - 6 | 1275 | 74 | |
| BT015893.1 | - | 5 - 6 | 1089 | 74 | ||
More Information
Set Membership: |
|

AT1G33360
Genomic Map