Assay Details
| Gene Symbol: AT1G33811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103101.4 | NP_564430.1 | 1 - 2 | 256 | 112 | |
| GenBank mRNA | AY099832.1 | - | 1 - 2 | 198 | 112 | |
| BT000322.1 | - | 1 - 2 | 146 | 112 | ||
More Information
Set Membership: |
|

AT1G33811
Genomic Map