Assay Details
| Gene Symbol: FAB1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333092.1 | NP_001319143.1 | 11 - 12 | 4140 | 66 | |
| NM_001333093.1 | NP_001321920.1 | 11 - 12 | 4070 | 66 | ||
| NM_001333094.1 | NP_001321919.1 | 7 - 8 | 3198 | 66 | ||
| NM_103148.4 | NP_174686.1 | 10 - 11 | 3883 | 66 | ||
More Information
Set Membership: |
|

FAB1D
Genomic Map