Assay Details
| Gene Symbol: FAB1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333092.1 | NP_001319143.1 | 10 - 11 | 2845 | 70 | |
| NM_001333093.1 | NP_001321920.1 | 10 - 11 | 2775 | 70 | ||
| NM_001333094.1 | NP_001321919.1 | 6 - 7 | 1903 | 70 | ||
| NM_103148.4 | NP_174686.1 | 9 - 10 | 2588 | 70 | ||
More Information
Set Membership: |
|

FAB1D
Genomic Map