Assay Details
| Gene Symbol: CYCB1;5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333104.1 | NP_001319145.1 | 8 - 9 | 1263 | 148 | |
| NM_001333105.1 | NP_001319146.1 | 8 - 9 | 1254 | 148 | ||
| NM_001333106.1 | NP_001320186.1 | 8 - 9 | 1263 | 148 | ||
| NM_001333107.1 | NP_001320188.1 | 8 - 9 | 1254 | 148 | ||
| NM_001333108.1 | NP_001320187.1 | 8 - 9 | 1263 | 148 | ||
| NM_001333109.1 | NP_001320190.1 | 8 - 9 | 1263 | 148 | ||
| NM_001333110.1 | NP_001320189.1 | 8 - 9 | 1263 | 148 | ||
| NM_001333111.1 | NP_001320191.1 | 7 - 8 | 1221 | 148 | ||
| NM_103168.4 | NP_564446.3 | 8 - 9 | 1263 | 148 | ||
More Information
Set Membership: |
|

CYCB1;5
Genomic Map